Learn More
BRAND™ LDPE Dropping Pipette
Product Code. 10717801 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1.5 mL
5 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10717801 1.5 mL Pack of 100
10604161 5 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10717801 Supplier BRAND™ Supplier No. 125410
  • ONLINE EXCLUSIVE PRICE
    367.00 NOK
    344.83 NOK / Pack of 100
    Save 22.17 NOK
    6% Off
Estimated Shipment: 30-11-2026
Low density polyethylene (LDPE) construction. Used for sampling and decanting infectious or toxic liquids. Graduated with bellows.
  • LDPE construction
  • Graduated
  • Includes bellows
  • For handling toxic or infections samples
  • Supplied in packs of 100
TRUSTED_SUSTAINABILITY
Specifications
Material LDPE
For Use With (Application) For Sampling and Decanting Infectious or Toxic Liquids
Graduated Yes
Quantity 100/Pk.
Volume (Metric) 1.5 mL
Length (Metric) 133 mm
Type Dropping Pipette