Learn More
Cole-Parmer™ Essentials Collapsible Containers, LDPE
LDPE Containers are collapsible when empty for easy storage.
650.00 NOK - 970.00 NOK
Specifications
Color | Translucent |
---|---|
For Use With (Application) | Containers nest uniformly when empty and expand readily when filled with liquid |
Material | LDPE |
Screw Color | White |
Closure Color | White |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11770039
|
Cole-Parmer™ Essentials
06100-10 |
1 L |
650.00 NOK
Pack of 12 |
Estimated Shipment: 07-11-2025
Log in to see stock. |
||||||
11779969
|
Cole-Parmer™ Essentials
06100-30 |
10 L |
870.00 NOK
Pack of 10 |
Estimated Shipment: 24-10-2025
Log in to see stock. |
||||||
11755519
|
Cole-Parmer™ Essentials
0610040 |
19 L |
970.00 NOK
Pack of 10 |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||||||
Description
- These containers nest uniformly when empty and expand readily when filled with liquid.
- They include polypropylene (PP) caps with injection-molded threads.
- Replacement caps sold separately.
Specifications
Translucent | |
LDPE | |
White | |
Collapsible Container | |
Screw Cap |
Containers nest uniformly when empty and expand readily when filled with liquid | |
White | |
Yes | |
Polypropylene |
Product Title
Cole-Parmer™ Essentials Collapsible Containers, LDPE