missing translation for 'onlineSavingsMsg'
Learn More

LDC000067, MedChemExpress

Product Code. p-200097244
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496888

Brand: Medchem Express HY15878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDC000067 is a highly specific CDK9 inhibitor with an IC50 value of 44±10 nM in vitro.

LDC000067 is a highly specific CDK9 inhibitor with an IC50 value of 44±10 nM in vitro.

Specifications

Chemical Name or Material LDC000067
CAS 1073485-20-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18N4O3S
Quantity 10 mg
Synonym LDC067
Solubility Information DMSO : ≥ 47 mg/mL (126.88 mM)
SMILES O=S(CC1=CC=CC(NC2=NC=NC(C3=CC=CC=C3OC)=C2)=C1)(N)=O
Molecular Weight (g/mol) 370.43
Formula Weight 370.43
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.