Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Tubing
- Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Product Code. 15288723
Change view
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
200 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
0.175 mm
0.2 mm
0.3 mm
0.53 mm
Unit Size:
Pack of 2
Pack of 5
| Product Code. | Overall Length (Metric) | Diameter (Metric) Inner | unitSize |
|---|---|---|---|
| 15288723 | 50 mm | 0.2 mm | Pack of 5 |
| 15258763 | 200 mm | 0.075 mm | Pack of 5 |
| 15258783 | 50 mm | 0.53 mm | Pack of 5 |
| 15268733 | 200 mm | 0.3 mm | Pack of 5 |
| 15268743 | 150 mm | 0.15 mm | Pack of 5 |
| 15268753 | 150 mm | 0.05 mm | Pack of 5 |
| 15268763 | 500 mm | 0.1 mm | Pack of 2 |
| 15298763 | 200 mm | 0.1 mm | Pack of 5 |
| 15248743 | 200 mm | 0.15 mm | Pack of 5 |
| 15298753 | 150 mm | 0.05 mm | Pack of 2 |
| 11991606 | 500 mm | 0.3 mm | Pack of 2 |
| 15208773 | 150 mm | 0.1 mm | Pack of 5 |
| 15258733 | 150 mm | 0.3 mm | Pack of 5 |
| 15268783 | 150 mm | 0.53 mm | Pack of 5 |
| 15238763 | 100 mm | 0.075 mm | Pack of 5 |
| 15278733 | 50 mm | 0.025 mm | Pack of 5 |
| 15298733 | 150 mm | 0.025 mm | Pack of 5 |
| 15208733 | 200 mm | 0.2 mm | Pack of 5 |
| 15208743 | 200 mm | 0.025 mm | Pack of 5 |
| 15218733 | 150 mm | 0.2 mm | Pack of 5 |
| 15218743 | 500 mm | 0.025 mm | Pack of 2 |
| 15228743 | 50 mm | 0.15 mm | Pack of 5 |
| 15228763 | 50 mm | 0.075 mm | Pack of 5 |
| 15238733 | 50 mm | 0.3 mm | Pack of 5 |
| 15238743 | 100 mm | 0.15 mm | Pack of 5 |
| 15238783 | 150 mm | 0.175 mm | Pack of 5 |
| 15248733 | 100 mm | 0.3 mm | Pack of 5 |
| 15248763 | 150 mm | 0.075 mm | Pack of 5 |
| 15248783 | 100 mm | 0.53 mm | Pack of 5 |
| 15278723 | 500 mm | 0.2 mm | Pack of 2 |
| 15278763 | 50 mm | 0.1 mm | Pack of 5 |
| 15278783 | 500 mm | 0.53 mm | Pack of 2 |
| 15288733 | 100 mm | 0.025 mm | Pack of 5 |
| 15288763 | 100 mm | 0.1 mm | Pack of 5 |
| 15298723 | 100 mm | 0.2 mm | Pack of 5 |
| 15258743 | 500 mm | 0.15 mm | Pack of 2 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
| Type | Tubing |
| Includes | 5/Pk. |
| Diameter (English) Outer | 1/16 in. |
| Diameter (Metric) Inner | 0.2 mm |
| Diameter (Metric) Outer | 1.6 mm |
| Overall Length (Metric) | 50 mm |
| Material | Polymer-sheathed Fused Silica |