Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Product Code. 15218733
Change view
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
200 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
0.175 mm
0.2 mm
0.3 mm
0.53 mm
Unit Size:
Pack of 2
Pack of 5
Product Code. Overall Length (Metric) Diameter (Metric) Inner unitSize
15218733 150 mm 0.2 mm Pack of 5
15258763 200 mm 0.075 mm Pack of 5
15258783 50 mm 0.53 mm Pack of 5
15268733 200 mm 0.3 mm Pack of 5
15268743 150 mm 0.15 mm Pack of 5
15268753 150 mm 0.05 mm Pack of 5
15268763 500 mm 0.1 mm Pack of 2
15298763 200 mm 0.1 mm Pack of 5
15248743 200 mm 0.15 mm Pack of 5
15298753 150 mm 0.05 mm Pack of 2
11991606 500 mm 0.3 mm Pack of 2
15208773 150 mm 0.1 mm Pack of 5
15258733 150 mm 0.3 mm Pack of 5
15268783 150 mm 0.53 mm Pack of 5
15238763 100 mm 0.075 mm Pack of 5
15278733 50 mm 0.025 mm Pack of 5
15298733 150 mm 0.025 mm Pack of 5
15208733 200 mm 0.2 mm Pack of 5
15208743 200 mm 0.025 mm Pack of 5
15218743 500 mm 0.025 mm Pack of 2
15228743 50 mm 0.15 mm Pack of 5
15228763 50 mm 0.075 mm Pack of 5
15238733 50 mm 0.3 mm Pack of 5
15238743 100 mm 0.15 mm Pack of 5
15238783 150 mm 0.175 mm Pack of 5
15248733 100 mm 0.3 mm Pack of 5
15248763 150 mm 0.075 mm Pack of 5
15248783 100 mm 0.53 mm Pack of 5
15278723 500 mm 0.2 mm Pack of 2
15278763 50 mm 0.1 mm Pack of 5
15278783 500 mm 0.53 mm Pack of 2
15288723 50 mm 0.2 mm Pack of 5
15288733 100 mm 0.025 mm Pack of 5
15288763 100 mm 0.1 mm Pack of 5
15298723 100 mm 0.2 mm Pack of 5
15258743 500 mm 0.15 mm Pack of 2
36 options
Product Code. 15218733
Brand: Trajan Scientific and Medical™ 0624205
Only null left
Add to basket
Add to basket
Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
    Specifications
    Type Tubing
    Includes 5/Pk.
    Diameter (Metric) Inner 0.2 mm
    Diameter (Metric) Outer 1.6 mm
    Overall Length (Metric) 150 mm
    Material Polymer-sheathed Fused Silica