Learn More
Lauric Acid Methyl Ester, Technical Grade, 99.1%, MP Biomedicals™
Product Code. 11491872 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11491872
Brand: MP Biomedicals 0210526325
Insoluble in water
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC
Specifications
Color Colorless
Assay Percent Range 99.1%
Quantity 25 g
Formula Weight 214.3
Grade Technical
Physical Form Liquid
Chemical Name or Material Lauric Acid Methyl Ester