Learn More
Kartell™ Latex Gas Sampling Balloon
Product Code. 10203763
Change view
Click to view available options
Capacity (Metric):
1 L
1.5 L
2 L
4 L
5 L
8 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10203763 2 L Each
10680374 1 L Each
10456123 1.5 L Each
10426503 4 L Each
10263183 5 L Each
10151183 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10203763 Supplier Kartell™ Supplier No. 0123200
  • ONLINE EXCLUSIVE PRICE
    257.00 NOK
    252.01 NOK / Pack of 1
    Save 4.99 NOK
    1% Off
View all promotions
In Stock
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Material Latex
Diameter (Metric) 160 mm
Product Type Gas Sampling Balloon
Capacity (Metric) 2 L