missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LASP1 Mouse anti-Human, Clone: 1G4B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68080-1-IG-20UL
This item is not returnable.
View return policy
Description
LASP1 Monoclonal antibody apecifically detetects LASP1 antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| LASP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| LASP1 | |
| LASP1 fusion protein Ag18101 | |
| 20 μL | |
| 3927 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG1 |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G4B6 | |
| Unconjugated | |
| BC012460 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction