Learn More
Brady™ LaserTab Laser Printable Polyolefin Labels
Product Code. 11513614
Change view
Click to view available options
Height (Metric):
6.4 mm
12.7 mm
21.2 mm
Labels per Row:
4
7
8
10
Width (Metric):
16.5 mm
20.3 mm
25.4 mm
47.6 mm
Width (Metric) Printable Area:
16.5 mm
20.3 mm
21.2 mm
25.4 mm
Unit Size:
1 pack
Product Code. Height (Metric) Labels per Row Width (Metric) Width (Metric) Printable Area unitSize
11513614 6.4 mm 8 20.3 mm 20.3 mm 1 pack
11503614 6.4 mm 10 16.5 mm 16.5 mm 1 pack
11523614 12.7 mm 7 25.4 mm 25.4 mm 1 pack
11533544 21.2 mm 4 47.6 mm 21.2 mm 1 pack
4 options
Product Code. 11513614
Brand: Brady™ 029760
Only null left
Add to basket
Add to basket
Blank, printable labels suitable for use with Laser Printers.
LaserTab Labels are available in the following formats:
  • Tamper-evident polyolefin
  • Polyester (UL Recognized Component when printed with designated laser printer toners)
  • Self-laminating
  • Nylon cloth
  • Paper
  • Polyimide
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyolefin (Tamper-evident)
Printable Sides Single-side Printable
Print Technology Laser
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Circuit Board Labeling, General Identification
For Use With (Equipment) Laser Printers
Horizontal Repeat (Metric) 25.4 mm
Labels per Row 8
Labels per Sheet 304
Min. Operating Temperature (Metric) -70°C
No. per Pack 10234 Labels/Pack
Quantity 10,234 Labels
Shape Rectangle
Temperature Range (English) -94°F to +212°F
Temperature Range (Metric) -70°C to +100°C
Thickness (English) 0.009 in.
Thickness (Metric) 0.23 mm
Dimensions (LxW) 6.35 x 20.32 mm
Length (English) 0.25 in.
Width (English) 0.8 in.
Width (Metric) 20.3 mm
Width (Metric) Printable Area 20.3 mm
Height (English) 0.25 in.
Height (English) Printable Area 0.25 in.
Height (Metric) 6.4 mm
Height (Metric) Printable Area 6.4 mm
Alternate Part Number LAT-4-410-10