Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Product Code. p-7077991
Change view
Click to view available options
Size (English):
0.25 x 0.25 in. (W x H)
0.3 x 0.25 in. (W x H)
0.375 x 0.375 in. (W x H)
0.5 x 0.437 in. (W x H)
0.65 x 0.2 in. (W x H)
0.65 x 0.25 in. (W x H)
0.8 x 0.25 in. (W x H)
0.8 x 0.5 in. (W x H)
1 x 0.5 in. (W x H)
1.5 x 0.25 in. (W x H)
1.875 x 0.375 in. (W x H)
1.875 x 0.833 in. (W x H)
1.875 x 1.33 in. (W x H)
2.5 x 1 in. (W x H)
3 x 0.375 in. (W x H)
8.5 x 11 in. (W x H)
Color:
White
Yellow
Unit Size:
1 pack
Product Code. Size (English) Color unitSize
11573584 1.875 x 0.375 in. (W x H) White 1 pack
11583584 3 x 0.375 in. (W x H) Yellow 1 pack
11503594 1.875 x 0.833 in. (W x H) White 1 pack
11503644 1.875 x 0.833 in. (W x H) Yellow 1 pack
11513594 1.875 x 1.33 in. (W x H) White 1 pack
11583564 0.3 x 0.25 in. (W x H) White 1 pack
11513644 2.5 x 1 in. (W x H) Yellow 1 pack
11533614 0.5 x 0.437 in. (W x H) Yellow 1 pack
11543614 0.25 x 0.25 in. (W x H) White 1 pack
11503574 0.375 x 0.375 in. (W x H) White 1 pack
11593614 8.5 x 11 in. (W x H) White 1 pack
11523644 8.5 x 11 in. (W x H) Yellow 1 pack
11523624 0.65 x 0.2 in. (W x H) White 1 pack
11523634 0.65 x 0.2 in. (W x H) Yellow 1 pack
11523574 0.65 x 0.25 in. (W x H) White 1 pack
11543574 0.8 x 0.25 in. (W x H) White 1 pack
11563574 0.8 x 0.5 in. (W x H) White 1 pack
11593574 1 x 0.5 in. (W x H) Yellow 1 pack
11553634 1 x 0.5 in. (W x H) White 1 pack
11533584 1.5 x 0.25 in. (W x H) White 1 pack
20 options
Product Code. 11573584
Brand: Brady™ 029730
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Certifications/Compliance UL Recognised; CSA Accepted
Description Chemical, high temperature, humidity, smear, UV and weather-Resistant
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester (B-747)
Printable Sides Single-side Printable
Print Technology Laser
Product Type Laser Printable Label
Sterility Non-sterile
For Use With (Application) Circuit board labelling, General identification
For Use With (Equipment) Laser Printers
Horizontal Repeat (Metric) 50.8 mm
Labels per Row 4
Min. Operating Temperature (Metric) -40°C
No. per Pack 5000 Labels/Pack
Quantity 5000/Pk.
Shape Rectangle
Size (English) 1.875 x 0.375 in. (W x H)
Size (Metric) 47.6 x 9.52 mm (W x H)
Temperature Range (English) -40°F to +266°F
Temperature Range (Metric) -40°C to +130°C
Thickness (English) 0.0037 in.
Thickness (Metric) 0.094 mm
Dimensions (LxW) 9.53 x 47.63 mm
Length (English) 1.875 in.
Width (English) 1.875 in.
Width (Metric) 47.63 mm
Width (Metric) Printable Area 47.63 mm
Height (English) 0.375 in.
Height (English) Printable Area 0.375 in.
Height (Metric) 9.53 mm
Height (Metric) Printable Area 9.53 mm
Alternate Part Number LAT-11-747-5