Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Product Code. 11503644
Change view
Click to view available options
Size (English):
0.25 x 0.25 in. (W x H)
0.3 x 0.25 in. (W x H)
0.375 x 0.375 in. (W x H)
0.5 x 0.437 in. (W x H)
0.65 x 0.2 in. (W x H)
0.65 x 0.25 in. (W x H)
0.8 x 0.25 in. (W x H)
0.8 x 0.5 in. (W x H)
1 x 0.5 in. (W x H)
1.5 x 0.25 in. (W x H)
1.875 x 0.375 in. (W x H)
1.875 x 0.833 in. (W x H)
1.875 x 1.33 in. (W x H)
2.5 x 1 in. (W x H)
3 x 0.375 in. (W x H)
8.5 x 11 in. (W x H)
Color:
White
Yellow
Unit Size:
1 pack
Product Code. Size (English) Color unitSize
11503644 1.875 x 0.833 in. (W x H) Yellow 1 pack
11573584 1.875 x 0.375 in. (W x H) White 1 pack
11583584 3 x 0.375 in. (W x H) Yellow 1 pack
11503594 1.875 x 0.833 in. (W x H) White 1 pack
11513594 1.875 x 1.33 in. (W x H) White 1 pack
11583564 0.3 x 0.25 in. (W x H) White 1 pack
11513644 2.5 x 1 in. (W x H) Yellow 1 pack
11533614 0.5 x 0.437 in. (W x H) Yellow 1 pack
11543614 0.25 x 0.25 in. (W x H) White 1 pack
11503574 0.375 x 0.375 in. (W x H) White 1 pack
11593614 8.5 x 11 in. (W x H) White 1 pack
11523644 8.5 x 11 in. (W x H) Yellow 1 pack
11523624 0.65 x 0.2 in. (W x H) White 1 pack
11523634 0.65 x 0.2 in. (W x H) Yellow 1 pack
11523574 0.65 x 0.25 in. (W x H) White 1 pack
11543574 0.8 x 0.25 in. (W x H) White 1 pack
11563574 0.8 x 0.5 in. (W x H) White 1 pack
11593574 1 x 0.5 in. (W x H) Yellow 1 pack
11553634 1 x 0.5 in. (W x H) White 1 pack
11533584 1.5 x 0.25 in. (W x H) White 1 pack
20 options
Product Code. 11503644
Brand: Brady™ 029794
Only null left
Add to basket
Add to basket
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Description Chemical, high temperature, humidity, smear, UV and weather-Resistant
Adhesive Type Acrylic, Permanent
Coating None
Color Yellow
Finish Matte
Format Label
Material Polyester (B-747)
Printable Sides Single-side Printable
Print Technology Laser
Product Type Laser Printable Label
Sterility Non-sterile
Quantity 2500/Pk.
Shape Rectangle
Size (English) 1.875 x 0.833 in. (W x H)
Size (Metric) 47.6 x 21.1 mm (W x H)
Thickness (English) 0.0037 in.
Dimensions (LxW) 21,16 x 47.63 mm
Length (English) 1.875 in.
Width (English) 1.875 in.
Width (Metric) 47.63 mm
Alternate Part Number LAT-13-747YL-2.5