Learn More
OHAUS™ Large Vessel Carrier
Product Code. 15732489 Shop All Ohaus Products
Change view
Click to view available options
Dimensions (L x W):
76.2 x 46 cm
61 x 61 cm
91 x 61 cm
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
15732489 61 x 61 cm Each
15722489 76.2 x 46 cm Each
15742489 91 x 61 cm Each
3 options
Product Code. 15732489
Brand: OHAUS™ 30400071
Only null left
Add to basket
Add to basket
OHAUS™ Large Vessel Carrier increases the amount of usable space for your samples.
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Tube racks and dimpled mats securely hold tubes in place
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vessel Carrier Platform
Material Stainless Steel
Depth (English) 24.00 in.
Depth (Metric) 61.00 cm
Dimensions (L x W) 61 x 61 cm
Length (English) 24.00 in.
Length (Metric) 61.00 cm
Width (English) 24.00 in.
Width (Metric) 61.00 cm
Description Large Vessel Carrier, 61 X 61 cm
Size Large