missing translation for 'onlineSavingsMsg'
Learn More

Laquinimod, MedChemExpress

Product Code. p-200101619
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
Product Code. Quantity unitSize
16455478 10 mg 10mg
16446066 100 mg 100mg
16421657 50 mg 50mg
16411656 500 mg 500mg
16408197 5 mg 5mg
5 options
This item is not returnable. View return policy

Product Code. 16455478

Brand: Medchem Express HY13010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Laquinimod (ABR-215062), an orally available carboxamide derivative, is a potent immunomodulator which prevents neurodegeneration and inflammation in the central nervous system.

Laquinimod reduces astrocytic NF-κB activation to protect from Cuprizone-induced demyelination. Laquinimod has the potential for relapsing remitting (RR) and chronic progressive (CP) forms of multiple sclerosis (MS; RRMS or CPMS) as well as neurodegenerative diseases research.

Specifications

Chemical Name or Material Laquinimod
Physical Form Solid
CAS 248281-84-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H17ClN2O3
For Use With (Application) COVID-19-immunoregulation
Synonym ABR-215062
Solubility Information DMSO : 100 mg/mL (280.27 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C2=C(N(C)C1=O)C=CC=C2Cl)N(CC)C3=CC=CC=C3
Molecular Weight (g/mol) 356.8
Formula Weight 356.8
Percent Purity 99.36%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.