missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-Biotin Antibody

Product Code. 10557774
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
Product Code. Quantity unitSize
10557774 25 μg Each
10308914 1 mg Each
2 options
This item is not returnable. View return policy

Product Code. 10557774

Brand: Thermo Scientific™ PV4745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ Tb-anti-Biotin Antibody

A general reagent for use in assay development, the anti-biotin antibody binds to biotinylated peptide or physiological protein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoprecipitation, Western Blot
Concentration 0.9mg/mL
Conjugate Tb (Terbium)/Lanthanide Chelates
Quantity 25 μg
Regulatory Status RUO
Research Discipline Target and Lead Identification and Validation
Primary or Secondary Primary
Packaging 1 Each
Includes 0.1% Sodium azide
Content And Storage Terbium-labeled anti-Biotin antibody in HEPES buffered saline. Store at -20°C.
Form Liquid
Excitation/Emission 340⁄490, 520
Shipping Condition Dry Ice
Readout End Point
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.