Learn More
Thermo Scientific™ LanthaScreen™ Eu-PY20 Antibody Kit
Description
- A general reagent to monitor tyrosine kinase activity
- The PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate
- For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications
Specifications
Specifications
| Conjugate | Europium/Lanthanide Chelates |
| Quantity | 1 mg |
| Content And Storage | Europium-labeled anti-PY20 antibody in HEPES buffered saline. Store at -20°C. |
| Detection Method | Fluorescence |
| Excitation/Emission | 340/615 |
| Shipping Condition | Dry Ice |
| Technique | TR-FRET |
| Validated Kinases | EGFR |
| Product Line | LanthaScreen |