Learn More
Lanoconazole, TRC
Specifications
Specifications
| Chemical Name or Material | Lanoconazole |
| CAS | 101530-10-3 |
| Molecular Formula | C14 H10 Cl N3 S2 |
| Quantity | 50 mg |
| Synonym | 1H-Imidazole-1-acetonitrile, α-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-, (E)-(±)-, (αE)-α-[4-(2-Chlorophenyl)-1,3-dithiolan-2-ylidene]-1H-imidazole-1-acetonitrile, 1H-Imidazole-1-acetonitrile, α-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-, (E)-, Astat, Latoconazole, NND 318, TJN 318 |
| InChI Formula | InChI=1S/C14H10ClN3S2/c15-11-4-2-1-3-10(11)13-8-19-14(20-13)12(7-16)18-6-5-17-9-18/h1-6,9,13H,8H2/b14-12+ |
| SMILES | Clc1ccccc1C2CS\C(=C(\C#N)/n3ccnc3)\S2 |
| IUPAC Name | (2E)-2-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-ylacetonitrile |
| Molecular Weight (g/mol) | 319.83 |
| Formula Weight | 319.0005 |