Learn More
Landiolol hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Landiolol hydrochloride |
| CAS | 144481-98-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H40ClN3O8 |
| Quantity | 5 mg |
| Synonym | ONO1101 hydrochloride |
| Solubility Information | DMSO : 250 mg/mL (457.83 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (183.13 mM) |
| SMILES | O=C(OC[C@H]1OC(C)(C)OC1)CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2.[H]Cl |
| Molecular Weight (g/mol) | 546.05 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.