Description
The LAMR1, RPSA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAMR1, RPSA. This antibody recognizes human, mouse, rat antigen. The LAMR1, RPSA antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | LAMR1 RPSA |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2A9A1 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 3921 |
| Show More |