Learn More
Fisherbrand™ Spare Lamp for Colorimeter
Shop All Fisherbrand Products
Click to view available options
Type:
Tungsten Halogen Lamp
Unit Size:
Each
Description
- Spare Tungsten Halogen lamp
- For use with Fisherbrand™ Digital Colorimeter
- Lamp life is extended as it is only on during the measurement cycle
- Easy to install
Specifications
Specifications
| Description | Spare lamp |
| Type | Tungsten Halogen Lamp |
| For Use With (Equipment) | Fisherbrand™ Digital Colorimeter |