Learn More
Sperian Protection™ Laminated Polypropylene With Polyethylene Film Mutex Light Overboots
Click to view available options
Size:
45
Unit Size:
100 pairs
Specifications
Specifications
| Color | White, Gray |
| For Use With (Application) | Work environments with low risk hazards : As benign cleaning products, diluted detergent solutions, fats, etc. |
| Height (Metric) | 57 mm |
| Length (Metric) | 32 mm |
| Material | Polypropylene, Polyethylene |
| Size | 45 |
| Product Type | Mutex Light Overboots |