Learn More
Brady™ Laminated Polyethylene Label
Brand: Brady™ 275019
Additional Details : Weight : 0.40000kg
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
1 Roll | |
Pipe markers and safety signs | |
46 mm | |
Label |
Polyethylene | |
Laminated Polyethylene | |
64 mm | |
DANGER environment |