Learn More
Brady™ Laminated Polyester Warning Signs
Product Code. 10775315
Click to view available options
Width (Metric):
50 mm
100 mm
Displayed Text:
Corrosive Batteries Warning Pictogram
Radioactive substances 7C III
Warning of Corrosive Substances
Warning of Hazardous Substances
Unit Size:
1 roll
Each
Product Code. 10775315
Brand: Brady™ 250210
Only null left
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 303-TRI 100-B7541 (BRANDED)
Material Polyester
Quantity 3/Pk.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of Corrosive Substances
Product Type Warning Sign
Color Black on Yellow