Learn More
Brady™ Laminated Polyester Warning Safety Signs
Product Code. 10041344
Change view
Click to view available options
Width (Metric):
100 mm
200 mm
Displayed Text:
Danger Zone Warning Pictogram - !
Warning about skid risk
Warning of industrial trucks
Warning of tripping hazard
Unit Size:
Each
Product Code. Width (Metric) Displayed Text unitSize
10041344 100 mm Danger Zone Warning Pictogram - ! Each
10151984 100 mm Warning about skid risk Each
10264894 200 mm Danger Zone Warning Pictogram - ! Each
10441074 100 mm Warning of industrial trucks Each
10061394 100 mm Warning of tripping hazard Each
5 options
Product Code. 10041344
Brand: Brady™ 250225
Only null left
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance
Alternate Part Number PIC 308-TRI 100-B7541 (BRANDED)
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Danger Zone Warning Pictogram - !
Product Type Warning Safety Sign
Color Black on Yellow