Learn More
- Home
- Products
- Facility Safety and Maintenance
- Signs and Tags
- Physical Hazard Signs
- Brady™ Laminated Polyester Transport Sign
Brady™ Laminated Polyester Transport Sign
Product Code. 11724894| Product Code. | Height (Metric) | Color | Displayed Text | Quantity | unitSize |
|---|---|---|---|---|---|
| 11724894 | 297 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 10725505 | 100 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 10254844 | 200 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11754884 | 297 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11740953 | 297 mm | Black on Red | Organic Peroxide 5.2 | 1 Ea. | Each |
| 10090284 | 100 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 10102414 | 100 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 10224894 | 100 mm | Black on White | Toxic and Harmful Substances 6.1 | 1 Ea. | Each |
| 11780993 | 100 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 11731003 | 100 mm | Black on White | Environmentally Hazardous Substances | 250/Roll | 1 roll |
| 10162454 | 200 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 10305964 | 200 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 10575884 | 200 mm | Black on White | Toxic and Harmful Substances 6.1 | 1 Ea. | Each |
| 11790993 | 200 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 11734894 | 297 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 11711003 | 297 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 10204514 | 100 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 10420884 | 200 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 10401844 | 100 mm | Black on Yellow | Warning of Toxic Substances | 3/Pk. | Each |
| 11704894 | 297 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 11700234 | 100 mm | Clear | Dangerous Substance 9 | 250/Roll | 1 roll |
| 11790224 | 100 mm | Clear | Corrosive Substance 8 | 250/Roll | 1 roll |
| 11780224 | 100 mm | Clear | Infectious Substance 6 | 250/Roll | 1 roll |
| 11770224 | 100 mm | Clear | Toxic Substance 6 | 250/Roll | 1 roll |
| 11700214 | 100 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11720214 | 297 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11750244 | 100 mm | White on Green | Non-flammable, non-toxic gas 2 | 250/Roll | 1 roll |
| 11710214 | 200 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
Product Code. 11724894
Brand: Brady™ 257584
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Certifications/Compliance | ISO 7010, RoHS compliant using EU Directive 2002/95/EC |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Pipe markers and safety signs |
| Height (Metric) | 297 mm |
| Thickness | 0.024 mm |
| Width (Metric) | 297 mm |
| Temperature Range (Metric) | -40°C to 110°C |
| Shape | Diamond |
| Displayed Text | Corrosive Substance 8 |
| Product Type | Transport Sign |
| Color | Black on White |
| Show More Show Less |