Learn More
Brady™ Laminated Polyester Transport Sign
Product Code. 11700234
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Color:
Black on Green
Black on Red
Black on White
Black on Yellow
Clear
White on Green
Displayed Text:
Corrosive Substance 8
Dangerous Substance 9
Environmentally Hazardous Substances
Infectious Substance 6
Miscellaneous Dangerous Substances and Articles 9
Non-flammable, non-toxic gas 2
Organic Peroxide 5.2
Oxidizing substances and organic peroxides 5.1
Toxic Substance 6
Toxic and Harmful Substances 6.1
Warning of Toxic Substances
Quantity:
1 Ea.
250/Roll
3/Pk.
Unit Size:
1 roll
Each
Product Code. 11700234
Brand: Brady™ 223586
Only null left
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number ADR 9RL-100*100-B7541
Finish Gloss
Material Polyester
Quantity 250/Roll
Adhesive Type Permanent
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Displayed Text Dangerous Substance 9
Product Type Transport Sign
Color Clear