Learn More
- Home
- Products
- Facility Safety and Maintenance
- Signs and Tags
- Physical Hazard Signs
- Brady™ Laminated Polyester Transport Sign
Brady™ Laminated Polyester Transport Sign
Product Code. 10725505
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Color:
Black on Green
Black on Red
Black on White
Black on Yellow
Clear
White on Green
Displayed Text:
Corrosive Substance 8
Dangerous Substance 9
Environmentally Hazardous Substances
Infectious Substance 6
Miscellaneous Dangerous Substances and Articles 9
Non-flammable, non-toxic gas 2
Organic Peroxide 5.2
Oxidizing substances and organic peroxides 5.1
Toxic Substance 6
Toxic and Harmful Substances 6.1
Warning of Toxic Substances
Quantity:
1 Ea.
250/Roll
3/Pk.
Unit Size:
1 roll
Each
28 product options available for selection
| Product Code. | Height (Metric) | Color | Displayed Text | Quantity | unitSize |
|---|---|---|---|---|---|
| 10725505 | 100 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 10254844 | 200 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11754884 | 297 mm | Black on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11740953 | 297 mm | Black on Red | Organic Peroxide 5.2 | 1 Ea. | Each |
| 10090284 | 100 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 10102414 | 100 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 10224894 | 100 mm | Black on White | Toxic and Harmful Substances 6.1 | 1 Ea. | Each |
| 11780993 | 100 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 11731003 | 100 mm | Black on White | Environmentally Hazardous Substances | 250/Roll | 1 roll |
| 10162454 | 200 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 10305964 | 200 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 10575884 | 200 mm | Black on White | Toxic and Harmful Substances 6.1 | 1 Ea. | Each |
| 11790993 | 200 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 11734894 | 297 mm | Black on White | Miscellaneous Dangerous Substances and Articles 9 | 1 Ea. | Each |
| 11724894 | 297 mm | Black on White | Corrosive Substance 8 | 1 Ea. | Each |
| 11711003 | 297 mm | Black on White | Environmentally Hazardous Substances | 1 Ea. | Each |
| 10204514 | 100 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 10420884 | 200 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 10401844 | 100 mm | Black on Yellow | Warning of Toxic Substances | 3/Pk. | Each |
| 11704894 | 297 mm | Black on Yellow | Oxidizing substances and organic peroxides 5.1 | 1 Ea. | Each |
| 11700234 | 100 mm | Clear | Dangerous Substance 9 | 250/Roll | 1 roll |
| 11790224 | 100 mm | Clear | Corrosive Substance 8 | 250/Roll | 1 roll |
| 11780224 | 100 mm | Clear | Infectious Substance 6 | 250/Roll | 1 roll |
| 11770224 | 100 mm | Clear | Toxic Substance 6 | 250/Roll | 1 roll |
| 11700214 | 100 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11720214 | 297 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
| 11750244 | 100 mm | White on Green | Non-flammable, non-toxic gas 2 | 250/Roll | 1 roll |
| 11710214 | 200 mm | White on Green | Non-flammable, non-toxic gas 2 | 1 Ea. | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
This item is not returnable.
View return policy
Product Code.
10725505
Supplier
Brady™
Supplier No.
257502
-
50.80 NOK
50.07 NOK / Pack of 1
Save 0.73 NOK
1% Off
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
48.26 NOK valid until 2026-08-14
BEST PRICE on Promo! Use promo code "28007" to get your promotional price.
BEST PRICE on Promo! Use promo code "28007" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Specifications
Specifications
| Certifications/Compliance | ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance |
| Alternate Part Number | ADR 2.2B-100*100-B7541 |
| Finish | Gloss |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Pipe markers and safety signs |
| Height (Metric) | 100 mm |
| Thickness | 0.024 mm |
| Width (Metric) | 100 mm |
| Temperature Range (Metric) | -40°C to 110°C |
| Shape | Rhomboid |
| Displayed Text | Non-flammable, non-toxic gas 2 |
| Product Type | Transport Sign |
| Color | Black on Green |
| Show More Show Less |