Learn More
Brady™ Laminated Polyester Transport Sign
Product Code. 10401844
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Color:
Black on Green
Black on Red
Black on White
Black on Yellow
Clear
White on Green
Displayed Text:
Corrosive Substance 8
Dangerous Substance 9
Environmentally Hazardous Substances
Infectious Substance 6
Miscellaneous Dangerous Substances and Articles 9
Non-flammable, non-toxic gas 2
Organic Peroxide 5.2
Oxidizing substances and organic peroxides 5.1
Toxic Substance 6
Toxic and Harmful Substances 6.1
Warning of Toxic Substances
Quantity:
1 Ea.
250/Roll
3/Pk.
Unit Size:
1 roll
Each
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Color Displayed Text Quantity unitSize
10401844 100 mm Black on Yellow Warning of Toxic Substances 3/Pk. Each
10725505 100 mm Black on Green Non-flammable, non-toxic gas 2 1 Ea. Each
10254844 200 mm Black on Green Non-flammable, non-toxic gas 2 1 Ea. Each
11754884 297 mm Black on Green Non-flammable, non-toxic gas 2 1 Ea. Each
11740953 297 mm Black on Red Organic Peroxide 5.2 1 Ea. Each
10090284 100 mm Black on White Corrosive Substance 8 1 Ea. Each
10102414 100 mm Black on White Miscellaneous Dangerous Substances and Articles 9 1 Ea. Each
10224894 100 mm Black on White Toxic and Harmful Substances 6.1 1 Ea. Each
11780993 100 mm Black on White Environmentally Hazardous Substances 1 Ea. Each
11731003 100 mm Black on White Environmentally Hazardous Substances 250/Roll 1 roll
10162454 200 mm Black on White Corrosive Substance 8 1 Ea. Each
10305964 200 mm Black on White Miscellaneous Dangerous Substances and Articles 9 1 Ea. Each
10575884 200 mm Black on White Toxic and Harmful Substances 6.1 1 Ea. Each
11790993 200 mm Black on White Environmentally Hazardous Substances 1 Ea. Each
11734894 297 mm Black on White Miscellaneous Dangerous Substances and Articles 9 1 Ea. Each
11724894 297 mm Black on White Corrosive Substance 8 1 Ea. Each
11711003 297 mm Black on White Environmentally Hazardous Substances 1 Ea. Each
10204514 100 mm Black on Yellow Oxidizing substances and organic peroxides 5.1 1 Ea. Each
10420884 200 mm Black on Yellow Oxidizing substances and organic peroxides 5.1 1 Ea. Each
11704894 297 mm Black on Yellow Oxidizing substances and organic peroxides 5.1 1 Ea. Each
11700234 100 mm Clear Dangerous Substance 9 250/Roll 1 roll
11790224 100 mm Clear Corrosive Substance 8 250/Roll 1 roll
11780224 100 mm Clear Infectious Substance 6 250/Roll 1 roll
11770224 100 mm Clear Toxic Substance 6 250/Roll 1 roll
11700214 100 mm White on Green Non-flammable, non-toxic gas 2 1 Ea. Each
11720214 297 mm White on Green Non-flammable, non-toxic gas 2 1 Ea. Each
11750244 100 mm White on Green Non-flammable, non-toxic gas 2 250/Roll 1 roll
11710214 200 mm White on Green Non-flammable, non-toxic gas 2 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Product Code. 10401844 Supplier Brady™ Supplier No. 250207
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 302-TRI 100-B7541 (BRANDED)
Material Polyester
Quantity 3/Pk.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of Toxic Substances
Product Type Transport Sign
Color Black on Yellow