Learn More
Brady™ Laminated Polyester Red and White Warning Panels
Product Code. 11736855 Shop All Brady Products
Change view
Click to view available options
Color:
Red on White
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11736855 Red on White 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11736855 Supplier Brady™ Supplier No. 255322
  • ONLINE EXCLUSIVE PRICE
    970.00 NOK
    953.03 NOK / Pack of 1
    Save 16.97 NOK
    1% Off
Estimated Shipment: 26-11-2026
These self-adhesive Warning Panels are designed to give early warning about trucks, loading platforms, posts and fork-lift trucks. They can be used wherever there are potential dangers to the work-force.
  • Designed to give early warning about trucks, loading platforms, posts and fork-lift trucks.
  • Can be used wherever there are potential dangers to the work-force.
Specifications
Alternate Part Number WP- 11X16 RD/WH
Certifications/Compliance RoHS compliant using EU Directive 2002/95/EC
Description Warning Panels - Red/White
Finish Gloss
For Use With (Application) Facility Identification
Height (English) 11.26 in.
Height (Metric) 286 mm
Packaging Quantity 10 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness 0.024 mm
Width (English) 16.85 in.
Width (Metric) 428 mm
Adhesive Type Permanent Acrylic
Color Red on White
Material Polyester
Type Warning Panels