Learn More
Brady™ Laminated Polyester Label Markers
Click to view available options
Color:
Black on Gray
White on Green
White on Purple
White on Red
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
Description
- Used for pipe markers and safety signs.
- B-7541 gives excellent adhesion to low surface energy surfaces, such as polypropylene and ABS, as well as on most powder coatings.
- Standard ink colors are black, red (RAL 3001-GL), yellow (RAL 1003-GL), blue (RAL5005-GL) and green (RAL 6032-GL).
Specifications
Specifications
| Description | Pipe Markers - Eau incendie |
| Adhesive Type | Permanent Acrylic |
| Color | White on Red |
| Finish | Gloss |
| Format | Roll |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Eau incendie |
| For Use With (Application) | Pipe and valve marking |
| Languages | French |
| Show More |