Learn More
Brady™ Laminated Polyester Label Markers
Product Code. 11705583
Click to view available options
Color:
Black on Gray
White on Green
White on Purple
White on Red
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
Product Code. 11705583
Brand: Brady™ 271388
Only null left
Add to basket
Add to basket
Laminated Polyester Label Markers feature a surface printed white polyester film with a permanent, acrylic based, pressure sensitive adhesive and over-laminated with a clear polyester film.
  • Used for pipe markers and safety signs.
  • B-7541 gives excellent adhesion to low surface energy surfaces, such as polypropylene and ABS, as well as on most powder coatings.
  • Standard ink colors are black, red (RAL 3001-GL), yellow (RAL 1003-GL), blue (RAL5005-GL) and green (RAL 6032-GL).
Specifications
Description Pipe Markers - Eau incendie
Adhesive Type Permanent Acrylic
Color White on Red
Finish Gloss
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Eau incendie
For Use With (Application) Pipe and valve marking
Languages French
No. per Pack 505 Markers/Roll
Quantity 505 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 33 m x 100 mm
Dimensions (L x W x H) 33 m x 100 mm x 60 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Height (English) 2.36 in.
Height (Metric) 60 mm
Height (Metric) Character 8 mm
Diameter (English) 3 in.
Diameter (Metric) 70 mm
Alternate Part Number PMF-EAUINCENDIE-RD3-RLL090