Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Use handrail
Product Code. p-7159857
Change view
Click to view available options
Diameter (Metric):
50 mm
100 mm
200 mm
315 mm
Unit Size:
Each
Pack of 2
Product Code. Diameter (Metric) unitSize
15614507 50 mm Pack of 2
15624507 100 mm Each
15306398 200 mm Each
15634507 315 mm Each
4 options
Product Code. 15614507
Brand: Brady™ 819610
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M012-Dia 050-PE-SHEET/2
Finish Gloss
Material Polyester
Quantity 2/Pk.
Adhesive Type Permanent Acrylic
Diameter (Metric) 50 mm
For Use With (Application) Indoor and Outdoor
Thickness 0.099 mm
Temperature Range (Metric) -40°C to 110°C
Shape Round
Languages Global
Color White on Blue