Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Evacuation assembly point
Product Code. p-7159801
Click to view available options
Width (Metric):
100 mm
148 mm
200 mm
250 mm
400 mm
Height (Metric):
100 mm
148 mm
200 mm
250 mm
400 mm
Unit Size:
Each
Product Code. 15604307
Brand: Brady™ 814913
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC E007-100x100-PE-CRD/1
Finish Gloss
Material Polyester
Adhesive Type Permanent Acrylic
Mounting Type Self-adhesive
For Use With (Application) Indoor and Outdoor
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Square
Languages Global
Color White on Green