Learn More
Brady™ Laminated Polyester Biohazard Warning Signs
Product Code. p-8004140
Change view
Click to view available options
Description:
Triangular, 100W x 87 mmH
Unit Size:
Each
Product Code. Description unitSize
10091344 Triangular, 100W x 87 mmH Each
1 options
Product Code. 10091344
Brand: Brady™ 250237
Only null left
Specifications
Alternate Part Number PIC 312-TRI 100-B7541 (BRANDED)
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Description Triangular, 100W x 87 mmH
Height (Metric) 87 mm
Width (Metric) 100 mm
Shape Triangular