Learn More
Brady™ Laminated Polyester Biohazard Warning Signs
Product Code. 10091344 Shop All Brady Products
Change view
Click to view available options
Description:
Triangular, 100W x 87 mmH
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10091344 Triangular, 100W x 87 mmH Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10091344 Supplier Brady™ Supplier No. 250237
  • ONLINE EXCLUSIVE PRICE
    106.00 NOK
    93.39 NOK / Pack of 1
    Save 12.61 NOK
    11% Off
View all promotions
In Stock
Specifications
Alternate Part Number PIC 312-TRI 100-B7541 (BRANDED)
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Description Triangular, 100W x 87 mmH
Height (Metric) 87 mm
Width (Metric) 100 mm
Shape Triangular