Learn More
Brady™ Laminated Polyester ADR 1.6 1 Explosive Substances Transport Signs
Product Code. 11720244
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Quantity:
1 Ea.
250/Pk.
Unit Size:
1 roll
Each
Product Code. Height (Metric) Width (Metric) Quantity unitSize
11720244 100 mm 100.00 mm 250/Pk. 1 roll
11770204 100 mm 100.00 mm 1 Ea. Each
11780204 200 mm 200.00 mm 1 Ea. Each
11790204 297 mm 297.00 mm 1 Ea. Each
4 options
Product Code. 11720244
Brand: Brady™ 223598
Only null left
Specifications
Alternate Part Number ADR 1.6RL-100*100-B7541
Finish Gloss
Material Laminated Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Displayed Text 1.6 1
Color Black on Orange