Description
Lamin B1 antibody apecifically detetects Lamin B1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.
Specifications
Specifications
| Antigen | Lamin B1 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Clone | 5H10 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | BC012295 |
| Gene Symbols | LMNB1 |
| Host Species | Rabbit |
| Immunogen | Lamin B1 fusion protein Ag3631 |
| Show More |