missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C Antibody (133A2) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97688JF635
This item is not returnable.
View return policy
Description
Lamin A + C Monoclonal antibody specifically detects Lamin A + C in Bovine,Canine,Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),CyTOF,Immunocytochemistry/ Immunofluorescence
Specifications
| Lamin A + C | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| Obtained from fusion of P3/X63.Ag8.653 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with partially purified recombinant human lamin A. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Protein Phosphatase, Signal Transduction, Stem Cells | |
| 4000 | |
| Store at 4°C in the dark. | |
| IgG3 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Immunocytochemistry/Immunofluorescence | |
| 133A2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Bovine, Canine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction