Learn More
Thermo Scientific™ Laemmli SDS Sample Buffer with Pyronin Y, non-reducing, 4X
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Pink |
| Concentration | 4X |
| Product Type | Sample Loading Buffer |
| Form | Liquid |
| For Use With (Application) | Laemmli SDS-PAGE |
| Quantity | 50 mL |