Learn More
LACTB2 Mouse anti-Human, Unconjugated, Clone: 1G6B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67399-1-IG-150UL
Description
The LACTB2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LACTB2. This antibody recognizes human, mouse antigen. The LACTB2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| LACTB2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1G6B1 | |
| Unconjugated | |
| 150 μL | |
| 51110 | |
| IgG1 |