Learn More
LACTB2 Mouse anti-Human, Unconjugated, Clone: 1G6B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67399-1-IG-20UL
 View more versions of this product
Product Code. 16850006
  • 1710.33 NOK / 20µL
Description
The LACTB2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LACTB2. This antibody recognizes human, mouse antigen. The LACTB2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
LACTB2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1G6B1
Unconjugated
20 μL
51110
IgG1