Learn More
Welch Ilmvac™ Labovac 10 Oil
Product Code. 11819580 Shop All Welch Ilmvac Products
Change view
Click to view available options
Volume (Metric):
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11819580 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11819580 Supplier Welch Ilmvac™ Supplier No. 800123
  • ONLINE EXCLUSIVE PRICE
    2560.00 NOK
    2429.36 NOK / 10 L
    Save 130.64 NOK
    5% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Use this mineral oil with standard rotary vane vacuum pumps. Welch Ilmvac™ Labovac 10 Oil is ideal for a variety of applications.
  • Mineral oil based vacuum oil for standard rotary vane vacuum pumps
  • 10L
  • For applications such as pumping out air, inert gases and water vapor, and also slightly acidic or basic vapors
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuum Oil
Volume (Metric) 10 L