Learn More
Juchheim Laborgeraete™ Laboratory Fire Suppression Trough
Click to view available options
Color:
Gray
Dimensions (L x W x H):
450 x 450 x 120 mm
Unit Size:
Each
Description
- Extinguishes burning liquids by removing oxygen
- Stainless steel tray and honeycomb insert
- With carrying handles
- Very good to clean
Specifications
Specifications
| Type | Storage Tub |
| Material | Stainless Steel |
| Color | Gray |
| Shape | Square |
| Dimensions (L x W x H) | 450 x 450 x 120 mm |