Learn More
Buerkle™ Laboratory Bottle PET Sterile
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Unit Size:
Each
Description
- Sterile SAL 10-3 (according to ISO 11137)
- PET crystal-clear
- With tamper-proof closure (HDPE)
- With moulded-in raised graduation
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | PET |