Learn More
Buerkle™ LaboPlast™ Bio Disposable Scoop
BIOPHARMAGREENER_CHOICE
Product Code. 15366327 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15366327 25 mL Pack of 10
15386327 100 mL Pack of 10
15376327 50 mL Pack of 10
15396327 150 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15366327 Supplier Buerkle™ Supplier No. 53790001
Produced in a Cleanroom Class 7 (10,000) and individually wrapped in bio-PE film for disposable use. Green PE bio-plastic has all the suitable properties of conventional PE but is manufactured from renewable raw materials.
  • Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled
  • The scoop stands horizontally as a result of the special handle design and the slightly angled front edge and can therefore be put down without the sample material trickling out.
  • Product and packaging made of renewable raw materials
  • Bio-PE, white
  • Individually wrapped
  • Production and packaging according to Cleanroom Class 7
  • With EU foodstuffs and FDA approval
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Sterility Non-sterile
Capacity (Metric) 25 mL
Color White
Certifications/Compliance FDA
Disposable Single-use
For Use With (Application) Foodstuffs
Material Polyethylene
Length (Metric) 141 mm