Learn More
Hirschmann™ Labopette™ Variable Volume Single Channel Manual Pipette
Description
- Variable volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
Specifications
| Accuracy | 0.8% |
| Certifications/Compliance | CE, IVD 98/79 EC |
| Compatible Tips | 1 to 200 μL |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Number of Channels | 1 |
| Product Line | Labopette |
| Type | Labopettor |
| Volume (Metric) | 10 to 100 μL |
| Includes | Calibration tool and pipette holder, individual serial number and calibration certificate |
| Show More |