Learn More
Hirschmann™ labopette™ fix
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
2000 μL
10 mL
Unit Size:
Each
Description
- Fixed volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
Specifications
| Accuracy | 0.8% |
| Certifications/Compliance | ISO 8655 |
| Compatible Tips | Clear: 100-5000 μL |
| Description | Tip ejector |
| Autoclavable | Autoclavable |
| Number of Channels | 1 |
| Type | Manual Pipette |
| Volume (Metric) | 2000 μL |
| Includes | Calibration device, individual number and weighing protocol |
| Precision | 0.3% |
| Show More |