Learn More
Hirschmann™ labopette™ fix
Product Code. 11602269
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
2000 μL
10 mL
Unit Size:
Each
Product Code. 11602269
Brand: Hirschmann™ 9475510
Ergonomically shaped manual pipette, effortless operation and tip ejector.
  • Fixed volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Accuracy 0.8%
Certifications/Compliance ISO 8655
Compatible Tips Blue: 10-1000 μL
Description Tip ejector
Autoclavable Autoclavable
Number of Channels 1
Type Manual Pipette
Volume (Metric) 1000 μL
Includes Calibration device, individual number and weighing protocol
Precision 0.3%
Quantity 1 Each