Learn More
Brady™ LabelMaker™ Vinyl Labels
Product Code. p-8001101
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
15113349 Each
15123349 Each
15103349 Each
3 options
Product Code. 15113349
Brand: Brady™ 131590
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LabelMaker™ Vinyl Labels are ideal for barcode labelling, circuit board labelling, component and equipment labelling, laboratory labelling, wire and cable labelling.
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant.
Specifications
Certifications/Compliance ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognised
Description BMP41/BMP51/BMP53 Labelmaker Labels
Adhesive Type Repositionable Rubber
Coating Vinyl
Color Black on White
Finish Semi-gloss
Format Label
Material Vinyl
Print Technology Thermal Transfer
Product Type Label
For Use With (Application) Barcode labelling, Circuit board labelling, Component and equipment labelling, Laboratory labelling, Wire and cable labelling
For Use With (Equipment) BMP41, BMP51, BMP53
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 160 Labels/Box
Quantity 160 Pk.
Shape Square
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness (English) 0.0073 in.
Thickness (Metric) 0.185 mm
Vertical Repeat (Metric) 28.58 mm
Wire Gauge 12 to 10 AWG
Dimensions (LxW) 25.4 x 25.4 mm
Length (English) 1 in.
Length (Metric) 25.4 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Diameter (English) 0.2 in.
Diameter (Metric) 5.1 mm