missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Label Dispenser for 150mm Wide Labels

Product Code. 15869481 Shop All Sampling Systems Products
Change view
Click to view available options
:
ProductTypeSpecs: Label Dispenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15869481 ProductTypeSpecs: Label Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869481 Supplier Sampling Systems™ Supplier No. 8220A17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sampling Systems™ Label Dispenser for 150mm Wide Labels is an easy way to store and dispense Ultra Adhesive Quality Control Labels.

  • Holds one roll of labels
  • Easy to use

Specifications

Product Type Label Dispenser
For Use With (Equipment) 150 x 150 mm wide labels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.