Learn More
Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Product Code. 10123292
Click to view available options
Height (Metric):
13.9 cm
14.4 cm
Unit Size:
Each
Product Code. 10123292
Brand: Labconco™ 7442600
Only null left
Add to basket
Add to basket
Holds tubes in upright position for freeze drying.
Hold tubes in upright position inside 900mL or larger Fast-Freeze Flasks and 750mL or larger Lyph-Lock Flasks Stainless steel
Specifications
Type Tube Holder
Capacity (Metric) 750, 900 mL
Material Stainless Steel
For Use With (Application) Hold tubes in upright position
For Use With (Equipment) 900ml or larger fast freeze™ flasks and 750ml or larger lyph lock flasks
Diameter (English) 3.5 in.
Diameter (Metric) 8.33 cm
Height (English) 5.6 in.
Height (Metric) 14.4 cm
Weight (English) 0.5 lb.
Weight (Metric) 0.5 kg